Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mig-22
Entrez Gene ID
176231
Gene Name
protein-coding
Gene Aliases
CELE_PAR2.4
Chromosome Location
Chr.III: 8763534 - 8767341 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001372637.3 NP_001360092.1 6 - 7 821 59
NM_066533.6 NP_498934.1 5 - 6 782 59
GenBank mRNA AB110823.1 - 4 - 5 845 59
AB110824.1 - 2 - 3 1123 59

View Full Product Details