Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
npp-9
Entrez Gene ID
175443
Gene Name
protein-coding
Gene Aliases
CELE_F59A2.1
Chromosome Location
Chr.III: 3399223 - 3404276 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001384053.1 7 - 8 2476 61
NM_001384054.1 7 - 8 2548 61

View Full Product Details