Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rol-8
Entrez Gene ID
174226
Gene Name
Cuticle collagen 6
Gene Aliases
CELE_ZK1290.3
Chromosome Location
Chr.II: 7544069 - 7545335 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063181.6 NP_495582.1 1 - 2 232 128

View Full Product Details