Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mnk-1
Entrez Gene ID
174623
Gene Name
protein-coding
Gene Aliases
CELE_R166.5
Chromosome Location
Chr.II: 10542421 - 10555051 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063871.6 NP_496272.2 5 - 6 1901 64
NM_182124.7 NP_871924.2 6 - 7 1851 64

View Full Product Details