Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pde-3
Entrez Gene ID
183981
Gene Name
protein-coding
Gene Aliases
CELE_E01F3.1
Chromosome Location
Chr.II: 14919705 - 14953353 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306640.4 NP_001293569.1 2 - 3 363 70
NM_001306642.3 NP_001293571.1 4 - 5 604 70
NM_001306643.3 NP_001293572.1 3 - 4 445 70
NM_001381668.1 NP_001367248.1 4 - 5 604 70
NM_001381670.1 NP_001368261.1 2 - 3 304 70
NM_001383996.1 4 - 5 668 70

View Full Product Details