Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lat-1
Entrez Gene ID
174419
Gene Name
protein-coding
Gene Aliases
CELE_B0457.1
Chromosome Location
Chr.II: 8896834 - 8908933 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001381541.1 NP_001367041.1 3 - 4 442 94
GenBank mRNA AY314771.1 - 3 - 4 442 94

View Full Product Details