Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pmt-1
Entrez Gene ID
173901
Gene Name
protein-coding
Gene Aliases
CELE_ZK622.3
Chromosome Location
Chr.II: 5278090 - 5282525 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383830.1 4 - 5 865 81
NM_062589.9 NP_494990.2 4 - 5 877 81
NM_062590.4 NP_494991.1 4 - 5 918 81
NR_002630.1 4 - 5 877 81

View Full Product Details