Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
btb-22
Entrez Gene ID
27219618
Gene Name
BTB domain-containing protein
Gene Aliases
CELE_F39E9.27
Chromosome Location
Chr.II: 3299757 - 3300698 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001322720.2 NP_001309553.1 3 - 3 545 69

View Full Product Details