Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- cks-1
- Entrez Gene ID
-
177658
- Gene Name
- Cyclin-dependent kinases regulatory subunit
- Gene Aliases
-
CELE_C09G4.3
- Chromosome Location
-
Chr.IV: 8511343 - 8514769 on Build WBcel235
- Gene Symbol
- C09G4.4
- Entrez Gene ID
-
3565561
- Gene Name
- Uncharacterized protein
- Gene Aliases
-
CELE_C09G4.4
- Chromosome Location
-
Chr.IV: 8511295 - 8513947 on Build WBcel235
- Gene Symbol
- mes-6
- Entrez Gene ID
-
177657
- Gene Name
- Polycomb protein mes-6
- Gene Aliases
-
CELE_C09G4.5
- Chromosome Location
-
Chr.IV: 8511284 - 8513921 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001383148.1 |
|
4 - 5 |
940 |
114 |
View Full Product Details