Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lntl-1
Entrez Gene ID
183098
Gene Name
protein-coding
Gene Aliases
CELE_C31H1.6
Chromosome Location
Chr.IV: 5800336 - 5807093 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001380241.1 NP_001367493.1 2 - 3 166 124
NM_068465.6 NP_500866.2 3 - 4 344 124
NR_131524.1 2 - 3 166 124

View Full Product Details