Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rol-3
Entrez Gene ID
179204
Gene Name
Protein roller-3
Gene Aliases
CELE_C16D9.2
Chromosome Location
Chr.V: 8228276 - 8238409 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001269168.3 NP_001256097.1 22 - 23 6127 101

View Full Product Details