Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
marc-6
Entrez Gene ID
172986
Gene Name
RING-CH-type domain-containing protein
Gene Aliases
CELE_F55A3.1
Chromosome Location
Chr.I: 10799623 - 10806934 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060422.6 NP_492823.2 2 - 3 268 163

View Full Product Details