Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
prp-17
Entrez Gene ID
172999
Gene Name
WD_REPEATS_REGION domain-containing protein
Gene Aliases
CELE_F49D11.1
Chromosome Location
Chr.I: 10936815 - 10940521 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060450.6 NP_492851.1 3 - 4 1627 70

View Full Product Details