Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
clec-93
Entrez Gene ID
191212
Gene Name
C-type lectin domain-containing protein
Gene Aliases
CELE_ZK39.4
Chromosome Location
Chr.I: 11142110 - 11144135 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060465.2 NP_492866.1 1 - 2 92 77

View Full Product Details