Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
clec-98
Entrez Gene ID
191215
Gene Name
C-type lectin domain-containing protein
Gene Aliases
CELE_ZK39.7
Chromosome Location
Chr.I: 11154213 - 11155369 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060470.4 NP_492871.2 3 - 4 291 72

View Full Product Details