Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sru-47
Entrez Gene ID
185345
Gene Name
Serpentine Receptor, class U
Gene Aliases
CELE_F36D1.3
Chromosome Location
Chr.I: 11289041 - 11291977 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060497.3 NP_492898.2 2 - 3 436 97

View Full Product Details