Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
png-1
Entrez Gene ID
173028
Gene Name
Peptide-N(4)-(N-acetyl-beta-glucosaminyl)asparagine amidase
Gene Aliases
CELE_F56G4.5
Chromosome Location
Chr.I: 11380231 - 11388751 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060512.5 NP_492913.1 8 - 9 1621 75

View Full Product Details