Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
aat-9
Entrez Gene ID
190250
Gene Name
protein-coding
Gene Aliases
CELE_Y53H1C.1
Chromosome Location
Chr.I: 11388681 - 11396662 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026617.4 NP_001021788.1 11 - 12 1526 72

View Full Product Details