Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
oac-19
Entrez Gene ID
173055
Gene Name
Acyl_transf_3 domain-containing protein
Gene Aliases
CELE_F35E2.6
Chromosome Location
Chr.I: 11707808 - 11712197 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060583.4 NP_492984.1 4 - 5 622 114

View Full Product Details