Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dnj-27
Entrez Gene ID
173065
Gene Name
protein-coding
Gene Aliases
CELE_Y47H9C.5
Chromosome Location
Chr.I: 11877803 - 11887278 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047239.4 NP_001040704.1 3 - 4 795 96

View Full Product Details