Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sdz-14
Entrez Gene ID
173101
Gene Name
SKN-1 Dependent Zygotic transcript
Gene Aliases
CELE_F14B6.1
Chromosome Location
Chr.I: 12216252 - 12217772 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060683.6 NP_493084.1 2 - 3 332 79

View Full Product Details