Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
oac-16
Entrez Gene ID
184447
Gene Name
Acyl_transf_3 domain-containing protein
Gene Aliases
CELE_F14B6.5
Chromosome Location
Chr.I: 12239160 - 12241808 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060688.2 NP_493089.2 7 - 8 1306 69

View Full Product Details