Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
oac-26
Entrez Gene ID
185602
Gene Name
Acyl_transf_3 domain-containing protein
Gene Aliases
CELE_F41D3.2
Chromosome Location
Chr.I: 12247771 - 12250210 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060690.1 NP_493091.1 6 - 7 1336 68

View Full Product Details