Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
oac-27
Entrez Gene ID
185604
Gene Name
Acyl_transf_3 domain-containing protein
Gene Aliases
CELE_F41D3.4
Chromosome Location
Chr.I: 12254485 - 12259455 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060692.3 NP_493093.1 9 - 10 1721 89

View Full Product Details