Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
oac-45
Entrez Gene ID
188335
Gene Name
Acyl_transf_3 domain-containing protein
Gene Aliases
CELE_T09E11.7
Chromosome Location
Chr.I: 12353506 - 12356337 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060726.1 NP_493127.1 9 - 10 1748 113

View Full Product Details