Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
oac-13
Entrez Gene ID
184022
Gene Name
Acyl_transf_3 domain-containing protein
Gene Aliases
CELE_E03H4.7
Chromosome Location
Chr.I: 12421239 - 12424411 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060749.2 NP_493150.2 7 - 8 1501 84

View Full Product Details