Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
clec-17
Entrez Gene ID
173111
Gene Name
C-type LECtin
Gene Aliases
CELE_E03H4.10
Chromosome Location
Chr.I: 12430028 - 12431500 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060751.6 NP_493152.1 1 - 2 248 71

View Full Product Details