Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sra-18
Entrez Gene ID
185054
Gene Name
Serpentine receptor class alpha-18
Gene Aliases
CELE_F28C12.2
Chromosome Location
Chr.I: 12692169 - 12693724 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060813.1 NP_493214.1 4 - 5 868 77

View Full Product Details