Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- bath-34
- Entrez Gene ID
-
24104971
- Gene Name
- BTB and MATH domain-containing protein 34
- Gene Aliases
-
CELE_W02A11.5
- Chromosome Location
-
Chr.I: 12750036 - 12752163 on Build WBcel235
- Gene Symbol
- bath-35
- Entrez Gene ID
-
24104973
- Gene Name
- BTB domain-containing protein
- Gene Aliases
-
CELE_W02A11.8
- Chromosome Location
-
Chr.I: 12750034 - 12759577 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001383738.1 |
|
2 - 3 |
588 |
66 |
View Full Product Details