Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ptr-17
Entrez Gene ID
173150
Gene Name
SSD domain-containing protein
Gene Aliases
CELE_Y18D10A.7
Chromosome Location
Chr.I: 12835115 - 12843335 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001377664.1 NP_001364498.1 5 - 6 1090 72

View Full Product Details