Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pad-1
Entrez Gene ID
173156
Gene Name
Protein pad-1
Gene Aliases
CELE_Y18D10A.13
Chromosome Location
Chr.I: 12878639 - 12902806 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060851.7 NP_493252.3 18 - 19 6156 107

View Full Product Details