Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nhr-85
Entrez Gene ID
191732
Gene Name
protein-coding
Gene Aliases
CELE_W05B5.3
Chromosome Location
Chr.I: 13055201 - 13062122 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026496.3 NP_001021667.1 1 - 2 299 82
GenBank mRNA AY204180.1 - 1 - 2 321 82

View Full Product Details