Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gcy-35
Entrez Gene ID
173202
Gene Name
protein-coding
Gene Aliases
CELE_T04D3.4
Chromosome Location
Chr.I: 13325230 - 13333513 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001265202.3 NP_001252131.1 13 - 14 1740 67
NM_001265204.3 NP_001252133.1 3 - 4 294 67
GenBank mRNA AY652944.1 - 12 - 13 1608 67

View Full Product Details