Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
trp-4
Entrez Gene ID
190574
Gene Name
ANK_REP_REGION domain-containing protein
Gene Aliases
CELE_Y71A12B.4
Chromosome Location
Chr.I: 13899085 - 13914008 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061028.3 NP_493429.2 2 - 3 367 67

View Full Product Details