Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
dnj-29
Entrez Gene ID
173278
Gene Name
protein-coding
Gene Aliases
CELE_Y63D3A.6
Chromosome Location
Chr.I: 14104425 - 14113108 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383758.1 6 - 7 1005 77
NM_061063.5 NP_493464.1 6 - 7 1005 77

View Full Product Details