Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pry-1
Entrez Gene ID
173287
Gene Name
Axin-like protein pry-1
Gene Aliases
CELE_C37A5.9
Chromosome Location
Chr.I: 14172824 - 14179360 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061073.6 NP_493474.2 4 - 5 388 61
GenBank mRNA AF468834.1 - 4 - 5 390 61

View Full Product Details