Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
eya-1
Entrez Gene ID
173293
Gene Name
protein-coding
Gene Aliases
CELE_C49A1.4
Chromosome Location
Chr.I: 14220693 - 14231360 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025884.6 NP_001021055.1 3 - 4 332 126
NM_001381258.2 NP_001367120.1 3 - 4 238 126

View Full Product Details