Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pes-7
Entrez Gene ID
173294
Gene Name
Patterned Expression Site
Gene Aliases
CELE_F09C3.1
Chromosome Location
Chr.I: 14263297 - 14271321 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061091.6 NP_493492.1 2 - 3 419 63

View Full Product Details