Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
zoo-1
Entrez Gene ID
173316
Gene Name
protein-coding
Gene Aliases
CELE_Y105E8A.26
Chromosome Location
Chr.I: 14529529 - 14546870 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001265447.3 NP_001252376.1 2 - 3 151 76
NM_001265448.4 NP_001252377.1 1 - 2 54 76
NM_001377679.2 NP_001364648.1 2 - 3 153 76

View Full Product Details