Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- lev-11
- Entrez Gene ID
-
173319
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_Y105E8B.1
- Chromosome Location
-
Chr.I: 14620814 - 14631254 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
D38542.1 |
- |
7 - 8 |
780 |
69 |
View Full Product Details