Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dnj-12
Entrez Gene ID
173341
Gene Name
DNaJ domain (prokaryotic heat shock protein)
Gene Aliases
CELE_F39B2.10
Chromosome Location
Chr.I: 14756858 - 14758790 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061169.7 NP_493570.1 3 - 4 758 112

View Full Product Details