Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
smp-1
Entrez Gene ID
173351
Gene Name
protein-coding
Gene Aliases
CELE_Y54E5B.1
Chromosome Location
Chr.I: 14807468 - 14815756 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383767.1 5 - 6 677 99
NM_061181.7 NP_493582.3 5 - 6 677 99

View Full Product Details