Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
unc-54
Entrez Gene ID
259839
Gene Name
Myosin-4
Gene Aliases
CELE_F11C3.3
Chromosome Location
Chr.I: 14855901 - 14863573 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061195.6 NP_493596.1 6 - 7 5363 74

View Full Product Details