Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ptr-23
Entrez Gene ID
173362
Gene Name
SSD domain-containing protein
Gene Aliases
CELE_ZK270.1
Chromosome Location
Chr.I: 14906792 - 14913082 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061202.6 NP_493603.1 3 - 4 672 80

View Full Product Details