Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fbxc-54
Entrez Gene ID
184901
Gene Name
F-box C protein
Gene Aliases
CELE_F23F1.3
Chromosome Location
Chr.II: 31719 - 33611 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061239.6 NP_493640.2 3 - 4 779 84

View Full Product Details