Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sptl-1
Entrez Gene ID
173389
Gene Name
protein-coding
Gene Aliases
CELE_C23H3.4
Chromosome Location
Chr.II: 51055 - 57587 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026808.4 NP_001021979.1 4 - 5 1220 77
NM_001313584.3 NP_001300513.1 1 - 2 440 77
NM_001383768.1 5 - 6 1374 77

View Full Product Details