Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cwn-1
Entrez Gene ID
173399
Gene Name
Protein Wnt-1
Gene Aliases
CELE_K10B4.6
Chromosome Location
Chr.II: 130983 - 133598 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061267.5 NP_493668.1 2 - 3 278 70
NR_132572.1 2 - 3 257 70
GenBank mRNA X72941.1 - - 272 70

View Full Product Details