Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ubxn-3
Entrez Gene ID
173407
Gene Name
protein-coding
Gene Aliases
CELE_F48A11.5
Chromosome Location
Chr.II: 216867 - 223785 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027010.5 NP_001022181.2 3 - 4 368 62
NM_001383772.1 5 - 6 481 62

View Full Product Details