Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sgcb-1
Entrez Gene ID
186816
Gene Name
SarcoGlyCan Beta homolog
Gene Aliases
CELE_K01A2.1
Chromosome Location
Chr.II: 326262 - 328224 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061309.5 NP_493710.2 2 - 3 240 112

View Full Product Details