Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ptr-3
Entrez Gene ID
173426
Gene Name
SSD domain-containing protein
Gene Aliases
CELE_C41D7.2
Chromosome Location
Chr.II: 362484 - 367553 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061317.7 NP_493718.2 4 - 5 529 76

View Full Product Details